Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWIST1 Mouse anti-Human, Mouse, Clone: 10E4E6, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Thermo Scientific MA538652

Encompass

Catalog No. PIMA538652



Description

Description

This antibody has been tested in indirect ELISA.

TWIST1 Monoclonal antibody specifically detects TWIST1 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

TWIST1
Monoclonal
1 mg/mL
PBS with 0.05% sodium azide
P26687, Q15672
AA960487; ACS3; B-HLH DNA binding protein; bHLHa38; BPES2; BPES3; charlie chaplin; class A basic helix-loop-helix protein 38; CRS; CRS1; CSO; H-twist; M-twist; p ACS3; Pde; pdt; Pluridigite; polydactyly EMS; SCS; Ska; Skam10Jus; Ska10; TWIST; twist basic helix-loop-helix transcription factor 1; twist family bHLH transcription factor 1; twist gene homolog 1; twist homolog 1; TWIST homolog of drosophila; Twist1; twist-related protein 1
Purified recombinant fragment of human TWIST1 (aa: 9-74) expressed in E. Coli.
Protein G
RUO
Antibody
Monoclonal
Human, Mouse
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
10E4E6
Unconjugated
TWIST1
Liquid, Purified
Mouse
IgG1
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
22160, 7291
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit