Learn More
Diversified Woodcrafts™ Two-Book Compartment Oak Table with ChemGuard Top: Electrical Option: 30 in. Height Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Ideal for middle school or for use with milder chemicals. Diversified Woodcraft Oak Tables with ChemGuard Tops incorporates the decorative features of high pressure laminate into a durable, chemical resistant surfacing material.
$977.00 - $1419.00
Specifications
Height (English) 30 in.
Plug Type US
Top Style 1.25 in. ChemGuard
Options Available UL rated electrical
Includes 2 books compartments and UL rated electrical
View More Specs
Products 15
Catalog Number Mfr. No. Depth (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Width (English) Price Quantity & Availability  
S38250 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7122K30E
30 in. 48 in. Each for $1,024.00
Add to cart
S38279 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7132K30E
30 in. 54 in. Each for $1,123.00
Add to cart
S38309 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7142K30E
30 in. 60 in. Each for $1,171.00
Add to cart
S38340 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7152K30E
30 in. 72 in. Each for $1,306.00
Add to cart
S38365 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7162K30E
21 in. 48 in. Each for $977.00
Add to cart
S38394 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7172K30E
36 in. 48 in. Each for $1,172.00
Add to cart
S38417 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7182K30E
21 in. 54 in. Each for $1,072.00
Add to cart
S47045 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7192K30E
36 in. 54 in. Each for $1,295.00
Add to cart
S47068 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7202K30E
24 in. 54 in. Each for $1,046.00
Add to cart
S47101 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7212K30E
21 in. 60 in. Each for $1,089.00
Add to cart
S47130 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7222K30E
36 in. 60 in. Each for $1,312.00
Add to cart
S47150 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7232K30E
21 in. 72 in. Each for $1,184.00
Add to cart
S47171 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7242K30E
36 in. 72 in. Each for $1,419.00
Add to cart
S47188 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7302K30E
24 in. 72 in. Each for $1,171.00
Add to cart
S47217 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7602K30E
24 in. 60 in. Each for $1,064.00
Add to cart
Description
Description
  • 1.25 in. ChemGuard Table Surface
  • UL rated electrical that can be hard wired or portable cord connected by certified electrician
  • 2 book compartments measuring 14W x 13.625D x 3.875 in. H
  • 2.25 in. solid square legs, with 5/16 in. hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16D x 4.5 in. H with heavy-duty 13 gauge steel corner braces
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Finished natural with a chemical resistant UV finish
  • Solid Oak Construction
  • Minimal assembly required; legs need to be attached
  • Weight capacity is 500lbs evenly distributed
  • SEFA compliant and MAS Certified Green
  • Limited Lifetime Warranty
  • Made in the USA
Order Info
Leg braces and stretchers are recommended for use with tables 72 in. wide or more. Leg braces and stretchers sold separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
30 in.
1.25 in. ChemGuard
2 books compartments and UL rated electrical
3.875 in.
Book Compartment Table
SEFA compliant and MAS Certified Green™
US
UL rated electrical
Wood (Red Oak)
14 in.
UV
No