Learn More
Diversified Woodcrafts™ Two-Book Compartment Oak Table with Solid Epoxy Top: 36 in. Height Products for Science Education Available on GSA/VA Contract for Federal Government customers only.
Ideal for high school and above with gas and other heat producting processes. Diversified Woodcraft Oak Table with Solid Epoxy Top is impervious to normal laboratory chemicals, heat and moisture.
$1311.00 - $1855.00
Specifications
Height (English) 36 in.
Top Style 1 in. Solid Epoxy
Includes 2-book compartments
Material Wood (Red Oak)
Height (English) Storage Area 3.875 in.
View More Specs
Products 10
Catalog Number Mfr. No. Depth (English) Width (English) Price Quantity & Availability  
Catalog Number Mfr. No. Depth (English) Width (English) Price Quantity & Availability  
S38220 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7106K36N
24 in. 48 in. Each for $1,365.00
Add to cart
S38270 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7126K36N
30 in. 48 in. Each for $1,559.00
Add to cart
S38299 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7136K36N
30 in. 54 in. Each for $1,745.00
Add to cart
S38331 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7146K36N
30 in. 60 in. Each for $1,855.00
Add to cart
S38385 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7166K36N
21 in. 48 in. Each for $1,311.00
Add to cart
S47036 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7186K36N
21 in. 54 in. Each for $1,411.00
Add to cart
S47090 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7206K36N
24 in. 54 in. Each for $1,456.00
Add to cart
S47121 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7216K36N
21 in. 60 in. Each for $1,514.00
Add to cart
S47164 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7236K34N
21 in. 72 in. Each for $1,720.00
Add to cart
S47239 Available on GSA/VA Contract for Federal Government customers only. Products for Science Education View Documents Diversified Woodcrafts™
C7606K36N
24 in. 60 in. Each for $1,591.00
Add to cart
Description
Description
  • 1 in. Epoxy Table Surface
  • 2 book compartments measuring 14W x 13.625D x 3.875 in. H
  • 2.25 in. solid square legs, with 5/16 in. hanger bolts installed, supplied nuts and washers
  • Aprons are 13/16D x 4.5 in. H with heavy-duty 13 gauge steel corner braces
  • Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
  • Finished natural with a chemical resistant UV finish
  • Solid Oak Construction
  • Minimal assembly required; legs need to be attached
  • Weight capacity is 500lbs evenly distributed
  • SEFA compliant and MAS Certified Green
  • Limited Lifetime Warranty
  • Made in the USA
Order Info
Leg braces and stretchers are recommended for use with tables 72 in. wide or more. Leg braces and stretchers sold separately.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
36 in.
2-book compartments
3.875 in.
Book Compartment Table
SEFA compliant and MAS Certified Green™
1 in. Solid Epoxy
Wood (Red Oak)
14 in.
UV
No