Learn More
Diversified Spaces™ Two-Book Compartment Oak Table with Solid Phenolic Top: Electrical Option: 30 in. Height

Ideal for high school and above with low temperature use. Diversified Spaces™ Oak Tables with Phelonic Tops are designed for use as counter tops, pegboards and reagent racks.
$862.00 - $1466.00
Specifications
Amperage | 15 A |
---|---|
Certifications | SEFA compliant and MAS Certified Green™ |
Finish | UV |
Frame Material | Red Oak |
Height (English) | 30 in. |
Catalog Number | Mfr. No. | Depth (English) | Width (English) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Depth (English) | Width (English) | Price | Quantity & Availability | ||||
S38203
![]() |
Diversified Spaces™
C7104K30E |
24 in. | 48 in. |
Each for $862.00
|
|
S38257
![]() |
Diversified Spaces™
C7124K30E |
30 in. | 48 in. |
Each for $1,008.00
|
|
S38286
![]() |
Diversified Spaces™
C7134K30E |
30 in. | 54 in. |
Each for $1,095.00
|
|
S38317
![]() |
Diversified Spaces™
C7144K30E |
30 in. | 60 in. |
Each for $1,172.00
|
|
S38346
![]() |
Diversified Spaces™
C7154K30E |
30 in. | 72 in. |
Each for $1,361.00
|
|
S38372
![]() |
Diversified Spaces™
C7164K30E |
21 in. | 48 in. |
Each for $871.00
|
|
S38401
![]() |
Diversified Spaces™
C7174K30E |
36 in. | 48 in. |
Each for $1,147.00
|
|
S38424
![]() |
Diversified Spaces™
C7184K30E |
21 in. | 54 in. |
Each for $912.00
|
|
S47052
![]() |
Diversified Spaces™
C7194K30E |
36 in. | 54 in. |
Each for $1,232.00
|
|
S47076
![]() |
Diversified Spaces™
C7204K30E |
24 in. | 54 in. |
Each for $929.00
|
|
S47108
![]() |
Diversified Spaces™
C7214K30E |
21 in. | 60 in. |
Each for $968.00
|
|
S47137
![]() |
Diversified Spaces™
C7224K30E |
36 in. | 60 in. |
Each for $1,295.00
|
|
S47155
![]() |
Diversified Spaces™
C7234K30E |
21 in. | 72 in. |
Each for $1,099.00
|
|
S47176
![]() |
Diversified Spaces™
C7244K30E |
36 in. | 72 in. |
Each for $1,466.00
|
|
S47194
![]() |
Diversified Spaces™
C7304K30E |
24 in. | 72 in. |
Each for $1,128.00
|
|
S47225
![]() |
Diversified Spaces™
C7604K30E |
24 in. | 60 in. |
Each for $1,012.00
|
|
Description
- 1 in. Phenolic Table Surface
- UL rated electrical that can be hard wired or portable cord connected by certified electrician
- 2 book compartments measuring 14W x 13.625D x 3.875 in. H
- 2.25 in. solid square legs, with 5/16 in. hanger bolts installed, supplied nuts and washers
- Aprons are 13/16D x 4.5 in. H with heavy-duty 13 gauge steel corner braces
- Equipped with non-skid adjustable glides for precise leveling and rubber leg boots
- Finished natural with a chemical resistant UV finish
- Solid Oak Construction
- Minimal assembly required; legs need to be attached
- Weight capacity is 500lbs evenly distributed
- SEFA compliant and MAS Certified Green
- Limited Lifetime Warranty
- Made in the USA
Order Info
Leg braces and stretchers are recommended for use with tables 72 in. wide or more. Leg braces and stretchers sold separately.
Specifications
15 A | |
UV | |
30 in. | |
Red Oak | |
UL rated electrical | |
125 V | |
14 in. | |
1 in. Solid Phenolic | |
Book Compartment Table |
SEFA compliant and MAS Certified Green™ | |
Red Oak | |
3.875 in. | |
US | |
1 Ea. | |
Lifetime | |
No | |
500 lb. | |
226.8 kg |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.