Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Brady™ Two-Sided Polyester Tag: TARGET ORGANS AFFECTED HEALTH HAZARD: FIRE HAZARD: REACTIVITY: SPECIFIC HAZARD: HAZARD RATINGS
Identify facility equipment to your exact specifications. Brady™ Two-Sided Polyester Tags are durable two-sided tags with a write-on area that accepts most marking pens.
Supplier: Brady™ 99102
Description
- Polyester tags provide identification solutions for almost any environment
- Metal grommet prevents tearing from the point of attachment
- Tag includes a write-on area that accepts most marking pens
- Tags are appropriate for indoor or outdoor use
- Tags are abrasion and chemical resistant
Specifications
25/Pk. | |
Black, Blue, Red, Yellow, White | |
TARGET ORGANS AFFECTED HEALTH HAZARD: FIRE HAZARD: REACTIVITY: SPECIFIC HAZARD: HAZARD RATINGS | |
Rectangle | |
3 x 5.75 in. |
OSHA 1910.145 | |
Polyester | |
(25) nylon fasteners | |
-40°F to 212°F | |
Not laminated |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction