Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Diversified Spaces™ Two-Student Single Storage Cabinet Table
Allows workspace and storage for two users.
Supplier: Diversified Spaces™ 2202K
Description
- Constructed from solid oak and hardwood veneers
- Features center cabinet with lock and rubber base molding
- Chemical-resistant, earth-friendly UV finish
Specifications
2-Student Single Storage Cabinet Table | |
24 x 66 x 30 in. | |
Plain apron; Two locking drawers | |
Red Oak, Hardwood Veneer |
ChemGuard top | |
UV | |
275 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction