Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TWS 119, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
1507095-58-0
C22H16F6N4O6
546.382
XJEFPMFFWJTJSL-UHFFFAOYSA-N
3-6-3-aminophenyl-7h-pyrrolo 2,3-d pyrimidin-4-yl oxyphenol ditrifluoroacetate
90488884
3-[[6-(3-aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]oxy]phenol;2,2,2-trifluoroacetic acid
C1=CC(=CC(=C1)N)C2=CC3=C(N2)N=CN=C3OC4=CC=CC(=C4)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O
This item is not returnable. View return policy
1507095-58-0
C22H16F6N4O6
546.382
XJEFPMFFWJTJSL-UHFFFAOYSA-N
3-6-3-aminophenyl-7h-pyrrolo 2,3-d pyrimidin-4-yl oxyphenol ditrifluoroacetate
90488884
3-[[6-(3-aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]oxy]phenol;2,2,2-trifluoroacetic acid
C1=CC(=CC(=C1)N)C2=CC3=C(N2)N=CN=C3OC4=CC=CC(=C4)O.C(=O)(C(F)(F)F)O.C(=O)(C(F)(F)F)O

GSK-3β inhibitor; induces neuronal differentiation in ESCs

Inhibitor of glycogen synthase kinase-3β (IC50 = 30 nM). Induces neuronal differentiation in pluripotent murine embryonal carcinoma cells and embryonic stem cells (ESCs).
Quantity 10mg
Formula Weight 546.38
Percent Purity >98%
Chemical Name or Material TWS 119

Recommended Storage : Desiccate at +4°C