Learn More
Thermo Scientific™ TX-750 Round Bucket Adapters Available on GSA/VA Contract for Federal Government customers only.
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters.
Manufacturer:  Thermo Scientific™ 75003699
 View more versions of this product
Catalog No. 75-003-699
  Check Availability
Add to cart
Specifications
Description
Add functionality to your rotor with Thermo Scientific™ TX-750 Round Bucket Adapters, which provide support for round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Adapters will hold round-bottom and conical tubes and bottles, microtubes and blood collection tubes.
Description & Specifications
Specifications
750mL Polypropylene Bio-Bottle (No Adapter Needed)
75003699