Learn More
Thermo Scientific™ TX-750 Swinging Bucket/BIOShield 1000A Adapters Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific? TX-750 Swinging Bucket/BIOShield? 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
Manufacturer:  Thermo Scientific™ 75003768
 View more versions of this product
Catalog No. 75-003-768
  Check Availability
Add to cart
Specifications
Description
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
  • Maximum tube size: 63 x 130mm
Description & Specifications
Specifications
5/7mL Blood Collection Tube