Learn More
Thermo Scientific™ TX-750 Swinging Bucket/BIOShield 1000A Adapters Available on GSA/VA Contract for Federal Government customers only.
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
$723.50 - $857.00
Products 8
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
75-003-737
View Documents
Thermo Scientific™
75003737
250mL Flat-bottom Bottles Pack of 4 for $841.00
Add to cart
75-003-755
View Documents
Thermo Scientific™
75003755
25mL Universal Tube Each for $768.00
Add to cart
75-003-756
View Documents
Thermo Scientific™
75003756
25mL DIN Round-bottom Tube Each for $768.00
Add to cart
75-003-758
View Documents
Thermo Scientific™
75003758
14mL Flanged Round-bottom Tube Each for $857.00
Add to cart
75-003-767 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
75003767
10mL Blood Collection/15mL DIN Tube Pack of 4 for $723.50
Add to cart
75-003-768 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
75003768
5/7mL Blood Collection Tube Pack of 4 for $723.50
Add to cart
75-003-769 Available on GSA/VA Contract for Federal Government customers only.
View Documents
Thermo Scientific™
75003769
5/7mL Round-bottom Tube Pack of 4 for $762.00
Add to cart
75-003-770
View Documents
Thermo Scientific™
75003770
1.5/2.0mL Conical Microtube Pack of 4 for $762.00
Add to cart
Description
Description
Use Thermo Scientific™ TX-750 Swinging Bucket/BIOShield™ 1000A Adapters to add versatility to TX-750/BIOShield Swinging-Bucket Rotors.
  • Adapters accept round/flat-bottom and blood collection bottles/tubes for TX-750 Rectangular Buckets or BIOShield 1000A Rotor.
  • Maximum tube size: 63 x 130mm
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.