Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TXLNG Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA551375
Description
Immunogen sequence: KALLQMAEEK TVRDKEYKAL QIKLERLEKL CRALQTERNE LNEKVEVLKE QVSIKAAIKA ANRDLATPVM QPCTALDSHK ELNTSSKRAL GAHLEAEPKS QRSAVQKPPS TGSAP Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 75%.
TXLNG gene ontology annotations related to this gene include regulation of bone mineralization; regulation of cell cycle process.
Specifications
TXLNG | |
Polyclonal | |
Unconjugated | |
TXLNG | |
4932441K18Rik; CXHXORF15; CXorf15; ELRG; environmental lipopolysaccharide-responding gene protein; environmental LPS-responding; factor inhibiting activating transcription factor 4 (ATF4)-mediated transcription; factor inhibiting ATF4-mediated transcription; FIAT; gamma-taxilin; lipopolysaccharide specific response-5 protein; lipopolysaccharide-responsive gene protein; Lipopolysaccharide-specific response protein 5; Lrg; Lrp; LSR5; taxilin gamma; TXLNG; TXLNGX | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55787 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NUQ3 | |
TXLNG | |
Recombinant protein corresponding to Human TXLNG. Recombinant protein control fragment (Product #RP-91990). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction