Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNDC11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP183994
Description
TXNDC11 Polyclonal specifically detects TXNDC11 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
TXNDC11 | |
Polyclonal | |
Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
EF-hand binding protein 1, EF-hand-binding protein 1, EFP1, thioredoxin domain containing 11, thioredoxin domain-containing protein 11 | |
Rabbit | |
Affinity Purified | |
RUO | |
51061 | |
Human | |
IgG |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
TXNDC11 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PSLNHIFIQLARNLPMDTFTVARIDVSQNDLPWEFMVDRLPTVLFFPCNRKDLSVKYPEDVPITLPNLLRFILHHSDPASSPQNVANSPTKECLQ | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction