Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNDC16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP19133920UL

 View more versions of this product

Catalog No. NBP19133920


Add to cart

Description

Description

TXNDC16 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

TXNDC16
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
TXNDC16
Synthetic peptide directed towards the N terminal of human TXNDC16. Peptide sequence EVAEDPQQVSTVHLQLGLPLVFIVSQQATYEADRRTAEWVAWRLLGKAGV.
Immunogen affinity purified
RUO
Primary
57544
Western Blot
Western Blot 0.2-1 ug/ml
NP_065835
KIAA1344, thioredoxin domain containing 16, thioredoxin domain-containing protein 16
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit