Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TXNDC6 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309902100UL
Description
TXNDC6 Polyclonal specifically detects TXNDC6 in Human samples. It is validated for Western Blot.Specifications
TXNDC6 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
MGC129586, NME9, thioredoxin domain containing 6, thioredoxin domain-containing protein 6, thioredoxin-like 2, Thioredoxin-like protein 2, TXL2, TXL-2 | |
The immunogen is a synthetic peptide directed towards the middle region of Human TXNDC6. Peptide sequence AEGRERKVIKDEALSDEDECVSHGKNNGEDEDMVSSERTCTLAIIKPDAV | |
100 μg | |
Protein Kinase | |
347736 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction