Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TXNL2 Mouse anti-Human, Mouse, Rat, Clone: 5G11, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA533226

Catalog No. PIMA533226



Description

Description

This gene encodes a member of the glutaredoxin family. Glutaredoxins are oxidoreductase enzymes that reduce a variety of substrates using glutathione as a cofactor. The encoded protein binds to and modulates the function of protein kinase C theta. The encoded protein may also inhibit apoptosis and play a role in cellular growth, and the expression of this gene may be a marker for cancer. Pseudogenes of this gene are located on the short arm of chromosomes 6 and 9. Alternatively spliced transcript variants have been observed for this gene.
Specifications

Specifications

TXNL2
Monoclonal
1 mg/mL
0.15M HEPES buffered saline with 0.01% BSA, 0.15M NaCl, 50% glycerol and 0.03% sodium azide
O76003, Q9CQM9
GLRX3
Recombit human protein purified from E.coli
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat
Immunocytochemistry
5G11
Unconjugated
GLRX3
Liquid
Mouse
IgG2b, kappa
100 μL
-20°C
Primary
10539, 30926
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit