Learn More
DuPont™ Tychem™ 2000 Tape
Keeps your PPE in place while providing the same level of chemical resistance as our Tychem™ 2000 garments.
Manufacturer:  DuPont™ QC0990YL000012NL
Catalog No. 19-088-900
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Tychem™ 2000 Tape keeps your PPE in place while providing the same level of chemical resistance as our Tychem™ 2000 garments
  • Helps to seal gaps in protective apparel ensemble closure points, in particular at seam, sleeve, glove, storm flap, and ankle/boot
  • Tears easily for quick application
  • Tape elasticity allows for tight adhesion to help prevent potential leak sources
  • Provides same chemical resistance as Tychem™ 2000 fabric. Provides protection against a variety of inorganic acids and bases
  • Flexibility and adhesive allows for easy repositioning
  • One size
Specifications
Specifications
Chemical
60 yds.