Learn More
DuPont™ Tychem™ 5000 Coverall with Respirator Hood, Berry Compliant
Click to view available options
Size:
Small
5X-Large
Description
DuPont™ Tychem™ 5000 helps offer protection for those in hazardous environments.
- Provides at least 30 minutes of protection against over 150 chemical challenges
- NFPA 1992 certified
Compliance
BN=BERRY COMPLIANT
Specifications
Size | 5X-Large |
Color | Tan |
Ankle Style | Attached Socks, Boot Overflaps |
Cuff Style | Elastic |
Closure Type | Storm Flap with Adhesive Closure |
Seam Type | Taped |
Style | Respirator Fit Hood |