Learn More
DuPont™ Tychem™ 5000 Coverall with Respirator Hood, NFPA 1992 Certified, Berry Compliant
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Description
DuPont™ Tychem™ 5000 helps offer protection for those in hazardous environments.
- Provides at least 30 minutes of protection against over 150 chemical challenges
- NFPA 1992 certified
Certifications
NFPA 1992
Compliance
BN=BERRY COMPLIANT
Specifications
Size | 2X-Large |
Color | Tan |
Ankle Style | Elastic |
Cuff Style | Elastic |
Closure Type | Double Storm Flap with Hook and Loop Seal |
Seam Type | Taped |
Certifications/Compliance | Certified to NFPA 1992 |
Style | Respirator Fit Hood |