Learn More
DuPont™ Tychem™ NP570 CT Gloves, Engineered Yarn Liner
Neoprene gloves provide chemical resistance against acids, caustics, solvents, greases and oils. Flexible and comfortable, these gloves also feature cut protection for a variety of applications.
Manufacturer:  DuPont™ NP5700BK100144EY
 View more versions of this product
Catalog No. 19-715-418
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Flexible coating with maximum comfort and protection
  • 13-gauge
  • Seamless
  • Cut-resistant
  • Engineered knit liner for maximum comfort and endurance
Specifications
Specifications
Black
Engineered Yarn
Neoprene
Tychem™ NP570 CT
14 in.
Cat III CE 2777, EN cut level 5
ANSI 5
No
AJKLMT
Specific
68 mil
Rough
Gauntlet
Lined
NP5700BK100144EY
10
35.6 cm
ANSI cut level A5
EN 388:3X43E
Type A
Bacteria, Fungi only
Yes
1.73 mm