Learn More
DuPont™ Tychem™ NP570 CT Glove
Provides ANSI level A5 and EN level 5 cut protection, as well as first-class defense against acids, caustics, solvents, greases and oils.
$804.61
Specifications
Certifications/Compliance ANSI cut level A5
Certifications/Compliance (Europe) EN cut level 5
Color Black
Disposable Yes
Exterior Finish Rough
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-715-416
View Documents
DuPont™
NP5700BK080144EY
8
Pack of 144 for $804.61
Add to cart
19-715-417
View Documents
DuPont™
NP5700BK090144EY
9
Pack of 144 for $804.61
Add to cart
19-715-418
View Documents
DuPont™
NP5700BK100144EY
10
Pack of 144 for $804.61
Add to cart
19-715-419
View Documents
DuPont™
NP5700BK110144EY
11
Pack of 144 for $804.61
Add to cart
Description
Description
  • Flexible coating with maximum comfort and protection
  • 13-gauge
  • Seamless
  • Cut-resistant
  • Engineered knit liner for maximum comfort and endurance
Specifications
Specifications
ANSI cut level A5
Black
Rough
Gauntlet
Yes
Engineered Yarn
Powder Free
68 mil
EN cut level 5
Yes
Black
Specific
14 in.
Neoprene
Non-sterile