Learn More
DuPont™ Tychem™ NT420 Glove
Provides chemical splash and oil protection
$402.00
Specifications
Color Blue
Disposable Yes
Exterior Finish Smooth
Glove Cuff Color Blue
Glove Cuff Style Rolled Cuff
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-715-381 View Documents DuPont™
NT4200BU061000UL
6 Pack of 1000 for $402.00
Add to cart
19-715-382 View Documents DuPont™
NT4200BU071000UL
7 Pack of 1000 for $402.00
Add to cart
19-715-383 View Documents DuPont™
NT4200BU081000UL
8 Pack of 1000 for $402.00
Add to cart
19-715-384 View Documents DuPont™
NT4200BU091000UL
9 Pack of 1000 for $402.00
Add to cart
Description
Description
  • Rubber free
  • Powder free
  • Smooth grip
Specifications
Specifications
Blue
Smooth
Rolled Cuff
Yes
Unlined
Powder-free
8 mil
Yes
Blue
Specific
9.5 in.
Nitrile
Non-sterile