Learn More
DuPont™ Tychem™ NT450 Glove
Flexible impermeable knit-lined dual nitrile glove. Ideal for heavy duty work or longer exposure to hazardous chemicals and solvents.
$463.50
Specifications
Color Blue
Disposable Yes
Exterior Finish Rough
Glove Cuff Color Blue
Glove Cuff Style Scalloped Edge
View More Specs
Products 4
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-715-391 View Documents DuPont™
NT4500BU080144KL
8 Pack of 144 for $463.50
Add to cart
19-715-392 View Documents DuPont™
NT4500BU090144KL
9 Pack of 144 for $463.50
Add to cart
19-715-393 View Documents DuPont™
NT4500BU100144KL
10 Pack of 144 for $463.50
Add to cart
19-715-394 View Documents DuPont™
NT4500BU110144KL
11 Pack of 144 for $463.50
Add to cart
Description
Description
  • Protection against an array of solvents, oils and acids.
  • Cotton liner absorbs perspiration for comfortable extended wear, with good mechanical resistance.
  • Protects the hand and forearm from oils, hydrocarbons, grease and abrasion.
  • Double nitrile coating provides excellent chemical, oil and abrasion resistance.
  • Impermeable for working in damp or greasy environments.
  • Rough textured finish over surface of hand.
Specifications
Specifications
Blue
Rough
Scalloped Edge
Yes
Cotton/Polyester
Powder-free
56 mil
Yes
Blue
Specific
14 in.
Nitrile
Non-sterile