Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DuPont™ Tychem™ NT480 Glove

Resistant to a range of solvents, animal fats and other chemicals

Manufacturer:  DuPont™ NT4800GR090288CF

 View more versions of this product

Catalog No. 19-715-404


Add to cart

Description

Description

  • Lightweight, bisque finish allows secure handling in wet and dry conditions.
  • Chlorinated
Specifications

Specifications

Safety
Rough
Specific
Cotton Flock
9
0.38 mm
Green
Green
330 mm
Yes
Gauntlet
Yes
Nitrile
15 mil
Powder-free
Non-sterile
13 in.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit