Learn More
DuPont™ Tychem™ 6000 FR Aprons
Click to view available options
Color:
Gray
Orange
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
Description
- Tychem™ 6000 FR garments are designed to combine the trusted chemical protection of Tychem™ and thermal/arc protection of Nomex into a single layer garment.
- Designed to help provide industrial workers and Hazmat responders with permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents and to provide escape time from flash fires and protection from electric arc.
- Fabric provides at least eight hours breakthrough protection against 17 of the 21 standard ASTM F1001 chemicals and at least 30 minutes of protection for more than 180 chemical challenges including chemical warfare agents.
- The arc rating of Tychem™ 6000 FR fabric is 15 cal/cm Ebt. This exceeds the NFPA 70E Hazard Risk Category 2 requirement of 8 cal/cm.
- Typical Applications:
- HazMat Teams in Fire Departments
- Industrial Fire Brigades
- Clandestine Lab Investigation
- Industrial Chemical Processing Plants
- Laboratories
- Tychem™ ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
- Sewn with Nomex thread
- Two buckle closure system
- Open back for wearer comfort
- Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
- Made in the USA
- North American Free Trade Agreement (NAFTA) compliant and Trade Agreement Act (TAA) compliant
Specifications
Color | Gray |
Closure Type | Buckles (Rear) |
Cuff Style | Elastic |
Seam Type | Taped |