Learn More
DuPont™ Tychem™ ThermoPro™ Sleeveless Gray Aprons
Designed to provide protection for industrial workers and Hazmat responders. DuPont™ Tychem™ ThermoPro™ Sleeveless Aprons provide permeation protection against a broad range of toxic industrial chemicals and chemical warfare agents
Manufacturer:  DuPont™ TP284TGYMD000400
 View more versions of this product
Catalog No. 19-122-001
Add to cart
Description
Description
  • Tychem™ ThermoPro for triple hazard protection
  • Sleeveless apron design provides frontal body protection
  • For flash fire and electric arc hazards, partial body protective Tychem ThermoPro garments must be used in conjunction with primary flame resistant clothing that is rated for the fire/arc hazard.
  • Sewn with Nomex™ thread
  • Designed with Nomex waist strap and flame resistant (FR) nylon buckle
  • Designed with end-user input for laboratory applications
  • Ideal for use on top of an existing flame resistant (FR) garment where extra chemical protection is required
  • Gray color for discretionary needs
  • Made in the USA
Order Info
Ansell™ Vinyl Closed-Back Aprons, Waist ties
Compliance
North American Free Trade Agreement (NAFTA) and Trade Agreement Act (TAA).
Specifications
Specifications
HazMat Teams in Fire Departments, Industrial Fire Brigades, Clandestine Lab Investigation, Industrial Chemical Processing Plants, Laboratories
Medium
Sleeveless
Gray
Taped