Learn More
Saint Gobain Performance Plastics Tube Tygon 7/16 x 9/16 50FT-15M/PK
Supplier: Saint Gobain Performance Plastics ACF00032
Saint-Gobain Tygon S3 E-3603 Flexible Tubings Diameter Inner 7/16 in. Diameter Outer 9/16 in. Maximum Pressure 13 psi at 73 deg.F Thickness 1/16 in. Clear