Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tylosin Tartrate, For Veterinary Use, BP, Spectrum™

Click to view available options
Quantity:
15 kg
25 kg
Packaging:
Fiber Drum
1405-54-5
C50H83NO23
1066.20
ICVKYYINQHWDLM-YLNALPTESA-N
(2R,3R)-2,3-dihydroxybutanedioic acid; 2-[(4R,5S,7R,9R,11E,13E,16R)-6-{[(2R,3R,4R,5S,6R)-5-{[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)C(O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(\C)=C\C1CO[C@@H]1O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]1OC
1405-54-5
C50H83NO23
1066.20
ICVKYYINQHWDLM-YLNALPTESA-N
(2R,3R)-2,3-dihydroxybutanedioic acid; 2-[(4R,5S,7R,9R,11E,13E,16R)-6-{[(2R,3R,4R,5S,6R)-5-{[(2S,4R,5S,6S)-4,5-dihydroxy-4,6-dimethyloxan-2-yl]oxy}-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-16-ethyl-4-hydroxy-15-({[(2R,3R,4R,5R,6R)-5-hydroxy-3,4-dimethoxy-6-methyloxan-2-yl]oxy}methyl)-5,9,13-trimethyl-2,10-dioxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
O[C@H]([C@@H](O)C(O)=O)C(O)=O.CC[C@H]1OC(=O)C[C@@H](O)[C@H](C)C(O[C@@H]2O[C@H](C)[C@@H](O[C@H]3C[C@@](C)(O)[C@@H](O)[C@H](C)O3)[C@@H]([C@H]2O)N(C)C)[C@@H](CC=O)C[C@@H](C)C(=O)\C=C\C(\C)=C\C1CO[C@@H]1O[C@H](C)[C@@H](O)[C@@H](OC)[C@H]1OC

T1483, 1405-54-5, (C46H77NO17)2•C4H6O6

Spectrum™ Tylosin Tartrate, For Veterinary Use, BP is a macrolide antibiotic used in veterinary medicine to treat bacterial infections in a wide range of species. Spectrum Chemical manufactured BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Fiber Drum
Loss on Drying 0.045
Quantity 15 kg
pH 5.0 to 7.2
CAS Max % 1
Grade BP