Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYMS Mouse anti-Human, Clone: UMAB227, liquid, UltraMAB™

Mouse Monoclonal Antibody
Supplier: OriGene UM800119

Description
Thymidylate Synthase (TS) (EC 2.1.1.45) converts deoxyuridine monophosphate (dUMP) to deoxythymidine monophosphate (dTMP), which is essential for DNA biosynthesis. TS is a critical target for the fluoropyrimidines, an important group of antineoplastic drugs that are widely used in the treatment of solid tumors. Both 5-FU and fluorodeoxyuridine are converted in tumor cells to FdUMP which inactivates TS by formation of a ternary covalent complex in the presence of the folate cofactor 5,10-methylenetetrahydrofolate. Expression of TS protein is associated with response to 5-fluorouracil (5-FU) in human colorectal, gastric, head and neck, and breast carcinomas.Specifications
TYMS | |
Monoclonal | |
Unconjugated | |
TYMS | |
HST422, TMS, TS | |
Mouse | |
Affinity Chromatography | |
RUO | |
7298 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
UMAB227 | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P04818 | |
TYMS | |
Full length human recombit protein of human TYMS (TS) produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title