Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3B Scientific™ Type II Diabetes Set
SDPSCI_ED

The 3B Scientific Type II Diabetes Set illustrates the effects associated with Type II Diabetes.

Manufacturer:  3B Scientific™ 1019573

Catalog No. S00269


Add to cart
This item is not returnable. View return policy

Description

Description

  • The Type II Diabetes Set includes miniature brain, eye, heart, kidney, artery, pancreas, neuron, and foot models.
  • Education card illustrates effects associated with Type II Diabetes: stroke, ocular pathology, hypertensive heart disease, hardening of the kidney, hardening of the arteries, insulin resistance, neuropathy, and foot ulcerations.
  • Models are reduced size
    • Model size: 10 in. Tall
    • Base: 8.8 x 6.25 in.
Specifications

Specifications

Type II Diabetes Set
10 in. Tall
2.43 lb.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit