Learn More
Typenex™ Blood Recipient Verification System
Ensures accuracy of blood cross-matching procedures
Manufacturer:  Typenex™ 4R4609
 View more versions of this product
Catalog No. 22-355-201
Add to cart
Includes: Each band supplied with 14 self-adhesive lables for up to 28 cross matches.
Description
Description
Self-contained, single-piece system establishes a positive physical link between the patient, the drawn blood sample, cross-matched units of blood and all paperwork, from bedside to lab to blood bank.
  • Durable Mylar™ wristbands are 22 in. to fit the wrist or angle of patients ranging from small children to large adults without the need for special extensions
  • Tamper-evident design resists removal until authorized
  • Self-contained cutter clip allows one-snap custom fitting; its smooth, flat clip design is comfortable against the skin
  • Easily removable labels are pre-printed with a unique alpha-numeric serialization to reduce cross-matching errors
  • Pre-printed full text prompts on sample tube help to reduce the number of redraws due to incomplete information
  • Waterproof bands and label will not smudge when wet
  • Arrows on tail show location of sticker release liner on back of tail
  • Made with nonallergenic, latex-free materials
Compliance
Meets American Hospital Association guidelines for a tamperproof, non-transferable wristband.
Specifications
Specifications
Self-cutting clip
Blood Band
Yes
One size fits all, adjustable length
Green
Designed specifically for transfusion patients and offers simple prompts for handwriting patient information
0.625 in.
1.58cm
Safety and Handling
Safety and Handling
ShelfLife : 72 hr.