Typenex™ Blood Recipient Verification System

Ensures accuracy of blood cross-matching procedures

Manufacturer: TYPENEX MEDICAL LLC 4R4608

 View more versions of this product

Catalog No. 22-355-200

  Check Availability
Add to Cart

    Specifications

    Color Red
    Product Type Blood Band
    For Use With (Application) Designed specifically for transfusion patients and offers simple prompts for handwriting patient information
    Latex Free Yes
    Width (English) 0.625 in.
    Size One size fits all, adjustable length
    Width (Metric) 1.58cm
    Shelf Life 72 hr.
    Closure Type Self-cutting clip

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit