Typenex™ Flexi Band Forms™

Provide perforated inserts for Flexi bands

Manufacturer: TYPENEX MEDICAL LLC FLX001

 View more versions of this product

Catalog No. 22-355-215

  Check Availability
Add to Cart

    Specifications

    Barcode Yes
    Product Type General Patient Identification Form
    For Use With (Equipment) Flexi bands
    Shelf Life 7+ Day(s)
    Includes 2 Patient inserts and 18 barcoded specimen tube stickers, 7-digit BBID in GS1-128 barcode format

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit