Learn More
Typenex™ Next Generation Bar Code Blood Band Handwriting Version
Accommodates and protects handwritten patient information
Manufacturer:  Typenex™ 4R4661
 View more versions of this product
Catalog No. 22-355-211
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Adhesive closure, no scissors required
  • Unique identifier (BBID: Blood Bank ID or armband number) in bar code and alphanumeric format
  • Fits most patient sizes
  • Compatible with R3 Attachment System to relocate, resize or replace on patient
  • One specimen tube sticker with unique BBID provides directions for proper overlabeling of the specimen tube with prompted patient information sticker
  • Two attachment adhesive tabs at the end of the detachable stickers affix to specimen for transport to blood bank
  • Unique BBID on specimen tube sticker can be scanned into LIS
  • Thirteen total detachable BBID stickers are available for paperwork, transfusion tags, donor unit labels, etc.
  • Four removable BBID stickers left on patient for use with the request for blood
  • Label donor units with unique BBID stickers from specimen
  • Ensure unique BBID matches on the recipient blood band, donor unit labels and any paperwork at the recipient bedside