Learn More
Typenex™ Original Blood Bands
Simple patient prompts for handwriting patient information
Manufacturer:  Typenex™ 4R4612
 View more versions of this product
Catalog No. 22-355-221
Add to cart
Description
Description
Intended for inpatient use up to 3 days
  • Adjustable length; closure with self-cutting clip
  • Bands are 0.625 in. wide
  • Latex-free
  • Available in blue and orange color
Specifications
Specifications
Self-cutting clip
Blood Band
Yes
Adjustable length
Orange
Designed specifically for transfusion patients and offers simple prompts for handwriting patient information
0.625 in.
1.58cm
Safety and Handling
Safety and Handling
ShelfLife : 72 hr.