Learn More
[Tyr0] Brain Natriuretic Peptide (1-32), Human, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66123MCR
 View more versions of this product
Catalog No. AAJ66123MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
3627.20
Human
Solid
RUO
[Tyr0] Brain Natriuretic Peptide (1-32), human
HPLC
1mg
[Tyr0] BNP (1-32), human
-30° to -10°C