Learn More
[Tyr4, D-Phe12] Bombesin, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66313LB0
 View more versions of this product
Catalog No. AAJ66313LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1679.89
5mg
Glp-Gln-Arg-Tyr-Gly-Asn-Gln-Trp-Ala-Val-Gly-D_Phe-Leu-Met-NH2
-30° to -10°C
[Tyr4, D-Phe12] Bombesin
HPLC
Solid
RUO
Bombesin Receptor Antagonist