Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
[Tyr0] Fibrinopeptide A, Human, Alfa Aesar
For cellular and molecular biology applications
Manufacturer: Alfa Aesar J66132LB0
Specifications
1699.73 | |
Human | |
Solid | |
RUO | |
[Tyr0] Fibrinopeptide A, human |
HPLC | |
5mg | |
Tyr-Ala-Asp-Ser-Gly-Glu-Gly-Asp-Phe-Leu-Ala-Glu-Gly-Gly-Gly-Val-Arg | |
-30° to -10°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only