Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYR Mouse anti-Human, Clone: OCA1/812, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB14623
Description
Mouse monoclonal antibody raised against full length recombinant human TYR.
- High specificity
- High reactivity
Specifications
TYR | |
Monoclonal | |
Unconjugated | |
ELISA Flow Cytometry (0.5-1 ug/million cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) The optimal working dilution should be determined by the end user. | |
P14679 | |
TYR | |
Recombinant protein corresponding to full length human TYR. | |
100 μg | |
Primary | |
Recognizes a cluster of proteins between 70-80 kDa, identified as TYR. Occasionally a minor band at 55 kDa is also detected. This antibody shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
OCA1/812 | |
Mouse monoclonal antibody raised against full length recombinant human TYR. | |
In 10 mM PBS. | |
OCA1A/OCAIA/SHEP3 | |
Mouse | |
Protein A/G purification | |
RUO | |
7299 | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction