Learn More
[Tyr8, Nle11] Substance P, Alfa Aesar™
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar J66679LB0
 View more versions of this product
Catalog No. AAJ66679LB0
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
1345.61
5mg
Arg-Pro-Lys-Pro-Gln-Gln-Phe-Tyr-Gly-Leu-Nle-NH2
-30° to -10°C
HPLC
Solid
RUO
[Tyr8, Nle11] Substance P