Learn More
Sklar InstrumentsSupplier Diversity Partner Sklar Black Tyrell Iris Hook, Sklar, 5 in., Single Hook Blunt, Stainless Steel with Reduced Reflectivity Polymer Coating. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 901056
Sklar Black Tyrell Iris Hook, Sklar, 5 in., Single Hook Blunt, Stainless Steel with Reduced Reflectivity Polymer Coating. The Sklar Black Tyrell Hook is commonly used in ophthalmic or dermatologic laser procedures. Sklar Black Laser instruments feature a non-reflective black polymer coating applied over the sandblasted instrument developed specifically for laser surgeries. This safe, durable coating reduces laser reflection to less than one-tenth of a percent, ensuring a safer surgical environment for patients and surgical staff. All Sklar Black coated instruments comply with biological testing requirements for cytotoxicity, sensitization regulatory, and ISO 10993 requirements. They also comply with the Pre-Vacuum and Gravity Cycle Steam Sterilization Validation of ANSI/AAMI ST79. Sklar Black laser coatings are guaranteed for one year against manufacturing defects. This product features a single blunt hook and it is 5 inches in length.
Catalog No. 50-143-6534