Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ Tyrosinase Ab-1, Mouse Monoclonal Antibody

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Ab-1, Mouse Monoclonal Antibody.

$207.50 - $1034.00

Specifications

Antigen Tyrosinase Ab-1
Clone T311
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Host Species Murine
View More Specs

Products 10
Catalog Number Mfr. No. Quantity Conjugate Format Formulation Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Format Formulation Price Quantity & Availability  
MS800B
View Documents
Thermo Scientific™
MS800B
1mL Biotin Concentrated with BSA and azide
Each for $945.00
Add to cart
 
MS800B0
View Documents
Thermo Scientific™
MS800B0
100μL Biotin Concentrated with BSA and azide
Each for $217.00
Add to cart
 
MS800B1
View Documents
Thermo Scientific™
MS800B1
500μL Biotin Concentrated with BSA and azide
Each for $737.00
Add to cart
 
MS800P
View Documents
Thermo Scientific™
MS800P
1mL Unconjugated Concentrated Purified with BSA and azide
Each for $1,034.00
Add to cart
 
MS800P0
View Documents
Thermo Scientific™
MS800P0
100μL Unconjugated Concentrated Purified with BSA and azide
Each for $207.50
Add to cart
 
MS800P1
View Documents
Thermo Scientific™
MS800P1
500μL Unconjugated Concentrated Purified with BSA and azide
Each for $455.00
Add to cart
 
MS800P1ABX
View Documents
Thermo Scientific™
MS800P1ABX
100μL Unconjugated Concentrated Purified without BSA and azide
Each for $495.00
Add to cart
 
MS800PABX
View Documents
Thermo Scientific™
MS800PABX
200μL Unconjugated Concentrated Purified without BSA and azide
Each for $808.00
Add to cart
 
MS800RQ
View Documents
Thermo Scientific™
MS800RQ
12mL Unconjugated Ready to Use
Each for $431.00
Add to cart
 
MS800R1
View Documents
Thermo Scientific™
MS800R1
1mL Unconjugated Prediluted Ready-to-use format This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Provide accurate, reproducible results with the Thermo Scientific™ Tyrosinase Ab-1, Mouse Monoclonal Antibody.

Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients.

Host Species: Mouse

Clone: T311

Isotype: IgG2a

Species Reactivity: Human, Dog and Cat. Weakly cross-reacts with Mouse. Others not known.

Epitope: aa 1-433

Immunogen: Recombinant tyrosinase protein

Molecular Weight: 70-80kDa

Positive Control: Melanoma cell lines and Melanoma

Cellular Localization: Cytoplasmic Ab-1 shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Staining of melanomas with Ab-1 showed tyrosinase in melanotic as well as amelanotic variants. Ab-1 is a useful marker for melanocytes and melanomas. Occasionally a minor band at 55kDa is also detected in Western Blotting

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunofluorescence
  • Immunocytology
Specifications

Specifications

Tyrosinase Ab-1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Murine
Cancer and Tumor Biology
Recombinant tyrosinase protein
T311
Monoclonal
IVD
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit