Learn More
Tyrosine Hydroxylase Mouse anti-Human, Mouse, Rat, Alexa Fluor 350, Clone: 5C7.2E8, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP242211AF350
Catalog No. NP242211A30
Add to cart
Description
Description
Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications
Specifications
Tyrosine Hydroxylase
5C7.2E8
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Mouse
IgG2a Kappa
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer
Primary
7054
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Alexa Fluor 350
50 mM Sodium Borate with 0.05% Sodium Azide
TH
This Tyrosine Hydroxylase Antibody (5C7.2E8) is made against a partial recombinant human Tyrosine Hydroxylase protein made to a C-terminal sequence (between amino acids 300-528) [Uniprot: P07101].
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Mouse, Rat