Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine Hydroxylase Mouse anti-Human, Rat, Alexa Fluor™ 532, Clone: 5C7.2E8, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP242211AF532

Catalog No. NP242211A32


Add to cart

Description

Description

Tyrosine Hydroxylase Monoclonal antibody specifically detects Tyrosine Hydroxylase in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin).
Specifications

Specifications

Tyrosine Hydroxylase
5C7.2E8
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Conjugated, Purified
TH
Partial recombinant human Tyrosine Hydroxylase protein made to a C-terminal sequence (between amino acids 300-528) [Uniprot: P07101]
Protein G purified
Cancer, Lipid and Metabolism, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Phospho Specific, Prostate Cancer
Primary
7054.0
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase
Mouse
IgG2a Kappa
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit