Learn More
AnaSpec TYROSINE KINASE PEPTIDE 1 5MG DFS Item
Manufacturer:  AnaSpec AS605515
Tyrosine Kinase Peptide 1 [KVEKIGEGTYGVVYK] 5mg. Peptide sequence KVEKIGEGTYGVVYK is derived from the amino acid residues CDC26-20. It is considered to be a generic substrate for various TPKs.
Catalog No. NC1474218