Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tyrosine, USP, 98.5-101.5%, Spectrum™

Click to view available options
Quantity:
25 g
100 g
1 kg
5 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
60-18-4
C9H11NO3
181.19
OUYCCCASQSFEME-UHFFFAOYNA-N
2-amino-3-(4-hydroxyphenyl)propanoic acid
NC(CC1=CC=C(O)C=C1)C(O)=O
60-18-4
C9H11NO3
181.19
OUYCCCASQSFEME-UHFFFAOYNA-N
2-amino-3-(4-hydroxyphenyl)propanoic acid
NC(CC1=CC=C(O)C=C1)C(O)=O

T1153, 60-18-4, C9H11NO3

Spectrum™ Tyrosine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Poly Bottle
Specific Rotation -9.8° to -11.2°
Loss on Drying 0.003
Ignition Residue 0.004
Quantity 1 kg
CAS Max % 1
Percent Purity 98.5 to 101.5%
Grade USP