Learn More
Aviva Systems Biology Corp TYRP1 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP44315_P050
This is a rabbit polyclonal antibody against TYRP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TYRP1. Alternate names: CAS2, CATB, GP75, TRP, TYRP, b-PROTEINApplicationIHC, WB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Sheep, Zebrafish
Catalog No. 50-014-53416