Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TYRP1 Mouse anti-Human, Clone: SPM611, Abnova™
Mouse monoclonal antibody raised against full length recombinant human TYRP1.
Supplier: Abnova Corporation MAB13172
Description
This gene encodes a melanosomal enzyme that belongs to the tyrosinase family and plays an important role in the melanin biosynthetic pathway. Defects in this gene are the cause of rufous oculocutaneous albinism and oculocutaneous albinism type III. [provided by RefSeq]
Specifications
TYRP1 | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (2-4 ug/mL) The optimal working dilution should be determined by the end user. | |
CAS2/CATB/GP75/TRP/TYRP/b-PROTEIN | |
Mouse | |
Protein A/G purification | |
RUO | |
7306 | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a κ |
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM611 | |
Mouse monoclonal antibody raised against full length recombinant human TYRP1. | |
In 10mM PBS. | |
TYRP1 | |
Recombinant protein corresponding to full length human TYRP1. | |
100 μg | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction