Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRP1 Rabbit anti-Human, Mouse, Allophycocyanin/Cy7, Clone: TYRP1/1564R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254446APCCY7

Catalog No. N254446APC7


Add to cart

Description

Description

TYRP1 Monoclonal antibody specifically detects TYRP1 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

TYRP1
TYRP1/1564R
Flow Cytometry
Conjugated, Purified
TYRP1
Recombinant human full-length TYRP1 protein
Protein A purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human, Mouse
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
b-PROTEIN, CAS25,6-dihydroxyindole-2-carboxylic acid oxidase, Catalase B, CATB, EC 1.14.18, EC 1.14.18.-, EC 1.14.18.1, Glycoprotein 75, GP75DHICA oxidase, Melanoma antigen gp75, TRP-1, TRPTYRP, tyrosinase-related protein 1TRP1, TYRRP
Rabbit
IgG Kappa
0.1 mL
Primary
7306.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit