Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TYRP1 Rabbit anti-Human, Unconjugated, Clone: TYRP1/2340R, Abnova™

Rabbit Monoclonal Antibody.

Manufacturer:  Abnova Corporation RAB00650

Catalog No. 89-339-665


Add to cart

Description

Description

TYRP1 Monoclonal antibody specifically detects TYRP1 in Human samples. It is validated for ELISA, IHC-P.
Specifications

Specifications

TYRP1
TYRP1/2340R
tyrosinase-related protein 1
Liquid
Rabbit
IgG
Store at -20 to -80°C
Monoclonal
Human
ELISA, Immunohistochemistry (Paraffin)
Unconjugated
TYRP1
CAS2, CATB, GP75, TRP, TYRP, b-PROTEIN
Recombinant protein corresponding to amino acids 257-377 of human TYRP1
100 μg
Primary
7306
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit