Tyrphostin B44, (-) enantiomer, Tocris

EGFR-kinase inhibitor

Manufacturer: R&D Systems 0578/50

Quantity 50mg

 View more versions of this product

Catalog No. 05-785-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >98%
    Molecular Formula C18H16N2O3
    Formula Weight 308.34
    Synonym (R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
    Chemical Name or Material Tyrphostin B44, (-) enantiomer

    Specifications

    Quantity 50mg
    Formula Weight 308.34
    Assay Percent Range >98%
    Molecular Formula C18H16N2O3
    CAS 133550-32-0
    Synonym (R)-(E)-2-Cyano-3-(3', 4'-dihydroxyphenyl)-N-(1-phenylethyl)-2-propenamide
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material Tyrphostin B44, (-) enantiomer

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit